missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-2-Aminoadipic acid, 98%, ACROS Organics™
$80.63 - $80.63
Chemical Identifiers
CAS | 1118-90-7 |
---|---|
Molecular Formula | C6H11NO4 |
Molecular Weight (g/mol) | 161.16 |
MDL Number | MFCD00002636 |
InChI Key | OYIFNHCXNCRBQI-BYPYZUCNSA-N |
Synonym | 2s-2-aminohexanedioic acid, aminoadipate, homoglutamic acid, l-2-amino adipic acid, l-2-aminoadipate, l-2-aminoadipic acid, l-alpha-aminoadipate, l-alpha-aminoadipic acid, l-homoglutamic acid, s-2-aminohexanedioic acid |
PubChem CID | 92136 |
ChEBI | CHEBI:37023 |
IUPAC Name | (2S)-2-aminohexanedioic acid |
SMILES | C(CC(C(=O)O)N)CC(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC302922500
|
Thermo Fisher Scientific
302922500 |
250mg | Glass bottle |
Each for $80.63
|
|
AC302920010
|
Thermo Fisher Scientific
AC302920010 |
1g | Glass bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1118-90-7 | |
161.16 | |
OYIFNHCXNCRBQI-BYPYZUCNSA-N | |
92136 | |
(2S)-2-aminohexanedioic acid |
C6H11NO4 | |
MFCD00002636 | |
2s-2-aminohexanedioic acid, aminoadipate, homoglutamic acid, l-2-amino adipic acid, l-2-aminoadipate, l-2-aminoadipic acid, l-alpha-aminoadipate, l-alpha-aminoadipic acid, l-homoglutamic acid, s-2-aminohexanedioic acid | |
CHEBI:37023 | |
C(CC(C(=O)O)N)CC(=O)O |
Specifications
Authentic | |
+ 22 | |
206°C | |
1118-90-7 | |
C6H11NO4 | |
MFCD00002636 | |
OYIFNHCXNCRBQI-BYPYZUCNSA-N | |
(2S)-2-aminohexanedioic acid | |
92136 | |
161.16 | |
98% |
Glass bottle | |
White | |
250mg | |
98% | |
HO2C(CH2)3CH(NH2)CO2H | |
2s-2-aminohexanedioic acid, aminoadipate, homoglutamic acid, l-2-amino adipic acid, l-2-aminoadipate, l-2-aminoadipic acid, l-alpha-aminoadipate, l-alpha-aminoadipic acid, l-homoglutamic acid, s-2-aminohexanedioic acid | |
C(CC(C(=O)O)N)CC(=O)O | |
161.16 | |
CHEBI:37023 | |
Solid | |
L-2-Aminoadipic acid, 98% |