Please login to your online account to display your discounted pricing

L-152,804, Tocris

Potent, selective non-peptide NPY Y5 receptor antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material L-152, 804
Assay Percent Range >99%
Molecular Formula C23H26O4
Formula Weight 366.46
Synonym 5,5-Dimethyl-2-(2,3,4,9-tetrahydro-3,3-dimethyl-1oxo-1H-xanthen-9-yl)-1,3-cyclohexanedione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

138250

 
r & d systems
RDS138250
50mg Each for $1,050.42

138210

 
r & d systems
RDS138210
10mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 366.46
Quantity 50mg
CAS 6508-43-6
Assay Percent Range >99%
Molecular Formula C23H26O4
Synonym 5,5-Dimethyl-2-(2,3,4,9-tetrahydro-3,3-dimethyl-1oxo-1H-xanthen-9-yl)-1,3-cyclohexanedione
Chemical Name or Material L-152, 804
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C