Please login to your online account to display your discounted pricing

L 006235, Tocris

Potent cathepsin K inhibitor

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material L 006235
Assay Percent Range >98%
Molecular Formula C24H30N6O2S
Formula Weight 466.6
Synonym N-[1-[[(Cyanomethyl)amino]carbonyl]cyclohexyl]-4-[2-(4-methyl-1-piperazinyl)-4-thiazolyl]benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

306610

 
r & d systems
RDS306610
10mg Each for $284.81

306650

 
r & d systems
RDS306650
50mg Each for $1,180.32
Description & Specifications

Specifications

Formula Weight 466.6
Quantity 10mg
Assay Percent Range >98%
Chemical Name or Material L 006235
Synonym N-[1-[[(Cyanomethyl)amino]carbonyl]cyclohexyl]-4-[2-(4-methyl-1-piperazinyl)-4-thiazolyl]benzamide
CAS 294623-49-7
Molecular Formula C24H30N6O2S
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C