Please login to your online account to display your discounted pricing

KY 02111, Tocris™

Promotes differentiation of human ESCs and iPSCs into cardiomyocytes

$177.37 - $714.82

Chemical Identifiers

Chemical Name or Material KY 02111
Assay Percent Range >99%
Molecular Formula C18H17ClN2O3S
Formula Weight 376.86
Synonym N-(6-Chloro-2-benzothiazolyl)-3, 4-dimethoxybenzenepropanamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

473110

 
r & d systems
RDS473110
10mg Each for $177.37

473150

 
r & d systems
RDS473150
50mg Each for $714.82
Description & Specifications

Specifications

Formula Weight 376.86
Quantity 10mg
Chemical Name or Material KY 02111
Recommended Storage Store at +4°C
Synonym N-(6-Chloro-2-benzothiazolyl)-3, 4-dimethoxybenzenepropanamide
Molecular Formula C18H17ClN2O3S
CAS 1118807-13-8
Assay Percent Range >99%
Safety and Handling

Store at +4°C