missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Epredia™ Lab Vision™ Ku (p70/p80) Ab-3 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Recommended for multiple molecular biology techniques, Epredia™ Ku (p70/p80) Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.

Supplier:  Epredia™ MS286P1

 View more versions of this product

Catalog No. MS286P1


Description

Description

The Ku autoantigen is a heterodimer of 70kDa (p70) and ∽80kDa (p80) proteins. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase that phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.

Host Species: Mouse

Clone: 162

Isotype: IgG2a

Species Reactivity: Human, Monkey, Xenopus. Mouse and Rat (Immunoprecipitation only). Does not react with cow and rabbit.

Epitope: A conformational epitope of p70/p80 dimer, which is destroyed during Western Blotting

Immunogen: Human B-cell nuclei from plasmacytoid 2p68 cells

Molecular Weight: 70kDa and 80kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Affinity Purification
  • Flow Cytometry
  • Gel Supershift
Specifications

Specifications

Ku (p70/p80) Ab-3
Monoclonal
200μg/mL
Mouse
500 μL
DNA Repair
Human, Non-human Primate, Xenopus species
IgG2a
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
162
Unconjugated
Human B cell nuclei from plasmacytoid 2p68 cells
RUO
Primary
Concentrated
SDS
Documents

Documents

Promotions

Promotions