Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Ku (p70/p80) Ab-3, Mouse Monoclonal Antibody


Recommended for multiple molecular biology techniques, Thermo Scientific™ Ku (p70/p80) Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  MS286P1ABX

 View more versions of this product

Catalog No. MS286P1ABX

  • / Each
 

Description & Specifications

Specifications

Isotype IgG2a
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Format Concentrated
Antigen Ku (p70/p80) Ab-3
Regulatory Status RUO
Clone 162
Cross Reactivity Human
Cross Reactivity Non-Human Primate
Cross Reactivity Xenopus
Quantity 100µL
Host Species Mouse
Immunogen Human B cell nuclei from plasmacytoid 2p68 cells
Type Primary
Conjugate Unlabeled
Formulation Ab without BSA/Azide (Frozen)
Research Discipline DNA Repair
Concentration 1mg/mL
Classification Monoclonal

Recommended for multiple molecular biology techniques, Thermo Scientific™ Ku (p70/p80) Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.

The Ku autoantigen is a heterodimer of 70kDa (p70) and ∽80kDa (p80) proteins. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase that phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.

Host Species: Mouse

Clone: 162

Isotype: IgG2a

Species Reactivity: Human, Monkey, Xenopus. Mouse and Rat (Immunoprecipitation only). Does not react with cow and rabbit.

Epitope: A conformational epitope of p70/p80 dimer, which is destroyed during Western Blotting

Immunogen: Human B-cell nuclei from plasmacytoid 2p68 cells

Molecular Weight: 70kDa and 80kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Affinity Purification
  • Flow Cytometry
  • Gel Supershift