missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kovac's Aldehyde Reagent, for Detection of Indole Producing Bacteria, Ricca Chemical

$216.55 - $806.55

Chemical Identifiers

CAS 71-36-3,7647-01-0,100-10-7
CAS Min % 63.0,30.35,5.15
CAS Max % 65.0,31.59,5.36
Physical Form Heavy Liquid
Chemical Name or Material Kovac's Aldehyde Reagent
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
4260-1 Ricca Chemical Company
RI42601
Glass amber 4L Each for $806.55
4260-16 SDS Ricca Chemical Company
RI426016
Glass amber 500mL Each for $216.55
Specifications

Description

Detection of Indole Producing Bacteria
Description & Specifications

Chemical Identifiers

71-36-3,7647-01-0,100-10-7
65.0,31.59,5.36
Kovac's Aldehyde Reagent
63.0,30.35,5.15
Heavy Liquid

Specifications

71-36-3,7647-01-0,100-10-7
65.0,31.59,5.36
Glass amber
4L
63.0,30.35,5.15
Heavy Liquid
Clear greenish-yellow
Kovac's Aldehyde Reagent