Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Vacuum Filtration Adapters


Use for reduced pressure filtration with plain stem Buchner funnels.

Manufacturer: kimble™  1781200014

 View more versions of this product

Catalog No. K17812-00014

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Fits Tubing ID (English) 0.25 in.
Fits Tubing ID (Metric) 6.35mm
For Use With (Application) Use for reduced pressure filtration with plain stem buchner funnels
For Use With (Equipment) Fits No. 2 pluro stopper
Height (English) 4.33 in.
Height (Metric) 110mm
Includes Hose connection, lower drip tip
Material Borosilicate Glass
Product Type Extended Body Vacuum Filtration Adapter
Standard Taper Joint No. 14/20

  • Top designed with a flange to accommodate a pluro stopper
  • Pluro stopper sold separately
  • Includes a lower drip tip
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements