Kontes Vacuum Adapters For Freeze Drying Rotary Evaporator Apparatus
Manufacturer:  Dwk Life Sciences Llc 5628500012
 View more versions of this product
Catalog No. K562850-0012
Specifications Description & Specifications
Specifications
0.31 in.
For connection to vacuum hose and a 562830 vacuum adapter
Viton™ o-ring
Hose Adapter
0.79cm
Vacuum hose and a 562830 vacuum adapter, freeze drying rotary evaporator apparatus
Polyethylene