Please login to your online account to display your discounted pricing
Kontes Vacuum Adapters For Freeze Drying Rotary Evaporator Apparatus
Manufacturer: kimble chase life science  5628300000
 View more versions of this product
Catalog No. K562830-0000
  • / Each
Description & Specifications
Specifications
Product Type Vacuum Adapter
Diameter (English) 0.5 in.
Diameter (Metric) 1.27cm
Includes Double viton™ o-ring seal, ptfe screen
Material Polyethylene
For Use With (Equipment) 562800 freeze dry flasks
For Use With (Application) For connection to vacuum hose and a 562830 vacuum adapter