Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Universal Inlet Adapters
Manufacturer: kimble™  1798002124
 View more versions of this product
Catalog No. K179800-2124
  • / Each
Description & Specifications
Specifications
Includes Viton O-rings and polypropylene cap
For Use With (Equipment) Design allows use with 675300 clamps
Product Type Inlet Adapter
Material PTFE
Standard Taper Joint No. 24/25