Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Universal Inlet Adapters
Manufacturer: kimble™  1798502324
 View more versions of this product
Catalog No. K179850-2324
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) For construct gas washing bottles
Includes Hose connection 2, O-rings are FKM, polypropylene compression cap
Product Type Universal Inlet Adapter
Standard Taper Joint No. 24/25
Material PTFE