Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Universal Adapters with Inlet
Use with pipets and syringes
Manufacturer: kimble™  1797502114
 View more versions of this product
Catalog No. K179750-2114
  • / Each
Description & Specifications
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
For Use With (Equipment) For plain thermometers, pipets, syringes or other small items with an O.D. from 2 to 11mm, 0.25 in. ID tubing
Includes Hose connection
Thread Size 13-425 GPI
Material Borosilicate Glass
Product Type Bevel-Seal™ Thermometer Inlet Adapter
Standard Taper Joint No. 14/20