Please login to your online account to display your discounted pricing
Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (90mm)
Designed for large volume filtration analysis of corrosive liquids or samples with high particulate loads that would tend to clog a 47 mm diameter filter
Manufacturer: kimble chase life science  9538250090
 View more versions of this product
Catalog No. K953825-0090
  • / Case
Description & Specifications
Specifications
Description Microfiltration Assembly
Material Glass
Capacity (Metric) 1L