missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm)

Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents

Manufacturer:  Kimble Chase Life Science 9538350000

 View more versions of this product

Catalog No. K953835-0000


 

Specifications Description & Specifications

Specifications

Microfiltration Assembly
Flask: 2000mL
Glass
Funnel: 500mL