DWK Life Sciences Kimble™ Kontes™ Fritted U-Shaped Purge and Trap Sparger
These U-shaped spargers are specially designed for use with Teledyne Tekmar™ 2000 and 3000 Purge and Trap Concentrators and with the 2016 and 2032 Autosamplers.
Supplier: DWK Life Sciences 5911012625
Description
- Fritted spargers are recommended for potable water and wastewater samples
- Options: 5mL Capacity with 3/4 in. mount or 25mL with 1/2 in. mount
- Case of 1
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
USP Type I, ASTM E438, Type I, Class A | |
Potable water and wastewater samples, soils, polymers, pharmaceuticals and foods | |
Borosilicate Glass | |
0.84 oz. |
25 mL | |
Teledyne Tekmar™ 2000 and 3000 purge and trap concentrators and with the 2016 and 2032 autosamplers | |
U |