missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kontes™ Tilting Repeating Dispensers

Fixed-volume models, from 5 to 100mL

Manufacturer:  Dwk Life Sciences Llc 7593000020

 View more versions of this product

Catalog No. K759300-0020


 

Specifications Description & Specifications

Specifications

33.81 oz., 1000mL
33.81 oz.
8.46 in.
29/42
11.41 in.
215mm
1000mL
Fixed volume dispensing head, Polyacetal standard taper clamp
Glass
20mL
290mm