missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Soxhlet/Dean-Stark Apparatus with Allihn Condenser

Avoid analyte loss and sample contamination

Manufacturer:  Kimble™ 5860000023

 View more versions of this product

Catalog No. K586000-0023


 

Requires : Complete assembly requires Cat. No. K586500-0023 or -0233 fritted glass extraction thimble (order separately online).

Specifications Description & Specifications

Specifications

1.92 in.
Avoid analyte loss and sample contamination
34cm
18cm
55/50
Soxhlet Extractor
4.9cm
13.38 in.
7.08 in.
24/40
Soxhlet/Dean-stark extraction apparatus