Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Serum Vials KG-33 Borosilicate Glass with Aluminum Seal


Manufacturer: kimble™  62121D10

 View more versions of this product

Catalog No. 10-310-242A

  • / Case of 864 Each
 

Description & Specifications

Specifications

Classification A
Volume (English) 2.75 dr.
Type Serum Vial
Material Borosilicate Glass
Certifications/Compliance ASTM E438, Type I, Class A
Volume (Metric) 10mL
Height (Metric) 54mm
Material (Seal) Aluminum
Color Clear
Capacity (Metric) 10.0mL
For Use With (Application) Freeze drying applications
Diameter (Metric) Outside 25mm