Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Schlenk Vacuum Tube with HI-VAC Valve
Suitable for use with K216050 or K216060 Manifolds
$268.79 - $401.21
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Product Type Vacuum Tube
For Use With (Equipment) 216050 or 216060 manifolds
Includes Hi-Vac™ valve, valve plug
View More Specs
Products
Catalog Number Mfr. No. Capacity (English) Height (English) Length (English) Width (English) Price Quantity    
K2187100015
kimble™
2187100015
0.5 oz. 3.93 in. 6.49 in. 2.55 in. Each for $268.79
K218710-0100
kimble™
2187100100
3.38 oz. 5.9 in. 5.9 in. 3.14 in. Each for $375.44
K218710-0350
kimble™
2187100350
11.83 oz. 9.84 in. 9.84 in. 3.54 in. Each for $401.21
K218710-0025
kimble™
2187100025
0.84 oz. 3.93 in. 6.49 in. 2.75 in. Each for $268.79
K218710-0050
kimble™
2187100050
1.69 oz. 4.72 in. 4.72 in. 2.95 in. Each for $314.05
Description & Specifications
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Product Type Vacuum Tube
For Use With (Equipment) 216050 or 216060 manifolds
Includes Hi-Vac™ valve, valve plug
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.