DWK Life Sciences Kimble™ Kontes™ Reflux Condensers with Coil Type and Joints
With an internal, coil-type cold finger; Tightly wound coil
Manufacturer:  DWK Life Sciences 4570000225
 View more versions of this product
Catalog No. 35-799
Description
Description
  • 24/40 joints
  • 175mm cold finger coil length
  • 170cm2 condensing area
  • No. 2 hose connection
Specifications
Specifications
USP Type I and ASTM E438, Type I, Class A
15.35 in.
Each
Borosilicate Glass
Condense high vapor pressure solvents like hexane
390 mm
24/40
Reflux Condenser