Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Reducing/Enlarging Connecting Adapters
Manufacturer: kimble™  1501000277
 View more versions of this product
Catalog No. K150100-0277
  • / Each
Description & Specifications
Specifications
For Use With (Application) For enlarging the size of Standard Taper glass joints
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Product Type Enlarging Connecting Adapter
Standard Taper Joint No. 45/50 Inner Bottom, 29/42 Outer Top