Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Reducing/Enlarging Connecting Adapters
$68.51 - $151.91
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Product Type Enlarging Connecting Adapter
For Use With (Application) For enlarging the size of Standard Taper glass joints
View More Specs
Products
Catalog Number Mfr. No. Standard Taper Joint No. Price Quantity    
K150100-0277
kimble™
1501000277
45/50 Inner Bottom, 29/42 Outer Top Each for $151.91
K274750-0241
kimble™
2747500241
19/22 Inner Bottom, 14/20 Outer Top Each for $68.51
K274750-0254
kimble™
2747500254
24/40 Inner Bottom, 19/22 Outer Top Each for $82.18
K274750-0242
kimble™
2747500242
24/40 Inner Bottom, 14/20 Outer Top Each for $79.49
K150100-0268
kimble™
1501000268
45/50 Inner Bottom, 24/40 Outer Top Each for $143.91
Description & Specifications
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Product Type Enlarging Connecting Adapter
For Use With (Application) For enlarging the size of Standard Taper glass joints
  • Outer joint is at the top, inner at the bottom
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class A requirements