DWK Life Sciences Kimble™ Kontes™ Reducing/Enlarging Connecting Adapters
Supplier: DWK Life Sciences 2747500254
Description
- Outer joint is at the top, inner at the bottom
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class A requirements
Specifications
USP Type I, ASTM E438, Type I, Class A | |
Borosilicate Glass | |
24/40 Inner Bottom, 19/22 Outer Top |
For enlarging the size of Standard Taper glass joints | |
Enlarging Connecting Adapter |