Kimble™ Kontes™ Reducing/Enlarging Connecting Adapters
Manufacturer:  Kimble™ 2747500241
 View more versions of this product
Catalog No. K274750-0241
Specifications Description & Specifications
Specifications
USP Type I, ASTM E438, Type I, Class A
Borosilicate Glass
19/22 Inner Bottom, 14/20 Outer Top
For enlarging the size of Standard Taper glass joints
Enlarging Connecting Adapter