Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Reducing Adapters
For connecting dissimilar standard taper joints
Manufacturer: kimble™  1501000261
 View more versions of this product
Catalog No. K150100-0261
  • / Each
Description & Specifications
Specifications
For Use With (Application) For connecting dissimilar joints
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Product Type Reducing Adapter
Standard Taper Joint No. 24/40 Outer Top, 10/30 Inner Bottom