DWK Life Sciences Kimble™ Kontes™ Reducing Adapters
For connecting dissimilar standard taper joints
$150.68 - $369.38
Specifications
Certifications/Compliance | USP Type I, ASTM E438, Type I, Class A |
---|---|
For Use With (Application) | For connecting dissimilar joints |
Material | Borosilicate Glass |
Product Type | Reducing Adapter |
Catalog Number | Mfr. No. | Standard Taper Joint No. | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Standard Taper Joint No. | Price | Quantity | |||||
K150100-0261
|
DWK Life Sciences
1501000261 |
24/40 Outer Top, 10/30 Inner Bottom |
Each for $186.24
|
|
|||||
K150100-0274
|
DWK Life Sciences
1501000274 |
24/40 Inner Bottom, 29/42 Outer Top |
Each for $223.88
|
|
|||||
K150100-0432
|
DWK Life Sciences
1501000432 |
24/40 Inner Bottom, 45/50 Outer Top |
Each for $369.38
|
|
|||||
K274750-0253
|
DWK Life Sciences
2747500253 |
19/22 Outer Top, 14/20 Inner Bottom |
Each for $150.68
|
|
|||||
K274750-0264
|
DWK Life Sciences
2747500264 |
24/40 Outer Top, 19/22 Inner Bottom |
Each for $188.65
|
|
|||||
K274750-0273
|
DWK Life Sciences
2747500273 |
19/22 Inner Bottom, 29/42 Outer Top |
Each for $196.74
|
|
|||||
Description
Outer joint at top; inner joint at bottom
Specifications
USP Type I, ASTM E438, Type I, Class A | |
For connecting dissimilar joints | |
Borosilicate Glass | |
Reducing Adapter |