DWK Life Sciences Kimble™ Kontes™ Reducing Adapters
For connecting dissimilar standard taper joints
Supplier: DWK Life Sciences 2747500273
Description
Outer joint at top; inner joint at bottom
Specifications
USP Type I, ASTM E438, Type I, Class A | |
Borosilicate Glass | |
19/22 Inner Bottom, 29/42 Outer Top |
For connecting dissimilar joints | |
Reducing Adapter |