Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Reaction-Jacketed Beakers


Manufacturer: kimble™  3170000500

 View more versions of this product

Catalog No. K317000-0500

  • / Each
 

Description & Specifications

Specifications

Material Borosilicate Glass
Product Type Beaker
Capacity (English) 16.9 oz.
Diameter (English) Outer 4.33 in.
Height (Metric) Outer 13cm
Capacity 16.9 oz., 500mL
Height (Metric) Inner 11.5cm
Certifications/Compliance ASTM E960, ASTM E438, USP Type I
Diameter (English) Inner 3.14 in.
Diameter (Metric) Inner 8cm
Connections 0.37 in. serrated hose
Diameter (Metric) Outer 11cm
Style Open Top
Height (English) Inner 4.52 in.
Height (English) Outer 5.11 in.
For Use With (Application) Laboratory applications requiring a variety of temperature-controlled reactions and useful in biological and chemical reactions including titrations, enzyme digestions and synthetic organic reactions
Includes Conical lower interior surface
Capacity (Metric) 500mL