missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Pressure Equalizing Lines, standard taper Stopcocks, Ground Joint Funnels

Manufacturer:  Kimble™ 6340000125

 View more versions of this product

Catalog No. K634000-0125


 

Specifications

Description

Funnels, addition; Kontes; PTFE-Plug pressure equalizing lines, stopcocks, ground joint funnels; Glass; standard taper No. (center): 24/40; Standard taper stopcock No.: 2; Capacity: 125mL

  • Glass stopcock
  • Standard taper joint: 24/40
  • Sold Individually

Description & Specifications

Specifications

4.22 oz.
Glass
24/40
2
4.22 oz., 125mL
345mm
USP Type I, ASTM E438 Type I
Pressure equalizing lines, stopcock
Addition Funnel
125mL
13.58 in.
Borosilicate Glass