Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Plug-Type Rubber Sleeve Stoppers

$124.67 - $127.94

Specifications

Color White
Material Rubber
Product Type Plug Type Sleeve Stopper
For Use With (Equipment) NMR tubes, pressure venting or adding reactants via syringe
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Outside Diameter (Metric) Outside Standard Taper Joint Taper Type Price Quantity    

K774261-0019

 
kimble™
7742610019
0.74 in. 1.9cm 19/22 19/22 Case of 50 Each for $124.67

K774261-0010

 
kimble™
7742610010
0.39 in. 1cm 10/18 10/18 Case of 50 Each for $127.94
Description & Specifications

Specifications

Color White
Material Rubber
Product Type Plug Type Sleeve Stopper
For Use With (Equipment) NMR tubes, pressure venting or adding reactants via syringe