Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Plug-Type Rubber Sleeve Stoppers


Manufacturer: kimble™  7742610019

 View more versions of this product

Catalog No. K774261-0019

  • / Case of 50 Each
 

Description & Specifications

Specifications

Diameter (English) Outside 0.74 in.
Color White
Material Rubber
Taper Type 19/22
Standard Taper Joint 19/22
Diameter (Metric) Outside 1.9cm
For Use With (Equipment) NMR tubes, pressure venting or adding reactants via syringe
Product Type Plug Type Sleeve Stopper