Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Pear-Shaped Flasks
Manufacturer: kimble™  2942600050
 View more versions of this product
Catalog No. K294260-0050
  • / Each
Description & Specifications
Specifications
Capacity (Metric) 50mL
Standard Taper Joint 19/22
Material Borosilicate Glass