DWK Life Sciences Kimble™ Kontes™ Pear-Shaped Heavy Wall Boiling Flask
Single-neck flasks with a Standard Taper outer joint at the top manufactured from 33 expansion, low extractable borosilicate glass.
Supplier:  DWK Life Sciences 6087000624
 View more versions of this product
Catalog No. K608700-0624
Description
Description
  • Available with a size 14/20, 19/22 or 24/40 taper joint
  • Heavy wall flasks in a range of capacities, from 5mL to 500mL
  • Conforms to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
500 mL
Borosilicate Glass
24/40