Kimble™ Kontes™ Oft Inlet Adapters
Manufacturer:  Kimble™ 1797102114
 View more versions of this product
Catalog No. K179710-2114
Specifications Description & Specifications
Specifications
One size 010 FKM O-ring and one 410119 cap
Offset Bevel-Seal™ Inlet Adapter
USP Type I, ASTM E438, Type I, Class A
Borosilicate Glass
14/20
13-425