Please login to your online account to display your discounted pricing

Kimble™ Kontes™ NMR Septum Sample Tube Caps


For 5mm, 7mm and 10mm NMR Tubes

Manufacturer: kimble™  8970970001

 View more versions of this product

Catalog No. K897097-0001

  • / Pack of 100 Each
 

Description & Specifications

Specifications

For Use With (Application) Provides penetration point for syringe or cannulation additions and useful as a closure or for sample outgassing
Material Rubber
Product Type Septa
For Use With (Equipment) 5mm thin-walled NMR sample tube for sample outgassing
Color White