Please login to your online account to display your discounted pricing

Kimble™ Kontes™ NMR Septum Sample Tube Caps


For 5mm, 7mm and 10mm NMR Tubes

Manufacturer: kimble™  8970980010

 View more versions of this product

Catalog No. K8970980010

  • / Case of 100 Each
 

Description & Specifications

Specifications

Color Red
Product Type Septa
For Use With (Application) Provides penetration point for syringe or cannulation additions and useful as a closure or for sample outgassing
For Use With (Equipment) 10mm thin-walled NMR sample tube for sample outgassing
Material Rubber