Please login to your online account to display your discounted pricing

Kimble™ Kontes™ NMR Sample Tube Caps


For Kontes™ 5 or 10mm NMR sample tubes

Manufacturer: kimble™  8970951011

 View more versions of this product

Catalog No. K897095-1011

  • / Case of 100 Each
 

Description & Specifications

Specifications

Color Assorted
Product Type Pressure Cap Variety Pack
For Use With (Application) Provides tight-fitting and stays in place even with a build-up of internal vaporization pressures
For Use With (Equipment) 10mm NMR sample tube
Includes 25 caps each of red, black, green, and yellow
Material Polyethylene