Kimble™ Kontes™ Inlet with Top Tubulature Adapters
$40.67 - $73.24
Specifications
Product Type Thermometer Inlet Adapter
For Use With (Equipment) For plain stem thermometers and gas inlet tubes
Includes Top tubulature, rubber adapter with a 0.25 in. hole
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Height (English) Height (Metric) Standard Taper Joint No. Price Quantity  
Catalog Number Mfr. No. Height (English) Height (Metric) Standard Taper Joint No. Price Quantity  
K199000-2440 Kimble™
1990002440
3.74 in. 95mm 24/40 Each for $73.24
K275410-1420 Kimble™
2754101420
1.96 in. 50mm 14/20 Each for $40.67
K275410-1922 Kimble™
2754101922
2.24 in. 57mm 19/22 Each for $44.78
Specifications Description & Specifications
Specifications
Thermometer Inlet Adapter
For plain stem thermometers and gas inlet tubes
Top tubulature, rubber adapter with a 0.25 in. hole