Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Inlet with Hose Connection Adapters


Manufacturer: kimble™  1810002440

 View more versions of this product

Catalog No. K181000-2440

  • / Each
 

Description & Specifications

Specifications

Fits Tubing ID (English) 0.37 in.
Fits Tubing ID (Metric) 9.39mm
For Use With (Equipment) For venting or introducing gas/vacuum into a system
Height (English) 13.97 in.
Height (Metric) 355mm
Includes Dual hose connections and standard taper joint
Material Borosilicate Glass
Standard Taper Joint No. 24/40
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Product Type Bleed Type Inlet Adapter