Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Inlet Flow Control Adapters
Manufacturer: kimble™  1850302942
 View more versions of this product
Catalog No. K185030-2942
  • / Each
Description & Specifications
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class AUSP Type I, ASTM E438, Type I, Class A
For Use With (Equipment) For regulating the input of gases or liquids into reaction systems
Height (English) 5.11 in.
Height (Metric) 130mm
Includes PTFE stopcock, hose connection 2
Material Borosilicate Glass
Product Type Right Angle Flow Control Inlet Adapter
Standard Taper Joint No. 29/42