Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Inlet Flow Control Adapters
$138.84 - $175.51
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class AUSP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Product Type Right Angle Flow Control Inlet Adapter
For Use With (Equipment) For regulating the input of gases or liquids into reaction systems
View More Specs
Products
Catalog Number Mfr. No. Height (English) Height (Metric) Includes Standard Taper Joint No. Price Quantity    
K185030-2942
kimble™
1850302942
5.11 in. 130mm PTFE stopcock, hose connection 2 29/42 Each for $166.80
K185000-2440
kimble™
1850002440
5.11 in. 130mm Glass stopcock and inner joint, hose connection 2 24/40 Each for $138.84
K275230-0000
kimble™
2752300000
3.54 in. 90mm PTFE stopcock, hose connection 1 14/20 Case of 20 Each for $147.05
K185030-2440
kimble™
1850302440
5.11 in. 130mm PTFE stopcock, hose connection 2 24/40 Case of 40 Each for $175.51
Description & Specifications
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class AUSP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Product Type Right Angle Flow Control Inlet Adapter
For Use With (Equipment) For regulating the input of gases or liquids into reaction systems