Kimble™ Kontes™ Inlet Adapters
Manufacturer:  Kimble™ 1860552440
 View more versions of this product
Catalog No. K18605-52440
Specifications Description & Specifications
Specifications
0.37 in.
For regulating the input of gases or liquids into reaction systems
130mm
Borosilicate Glass
24/40
9.39mm
5.11 in.
PTFE stopcock
Straight Flow Control Inlet Adapter