Please login to your online account to display your discounted pricing

Kimble™ Kontes™ HPLC Ultra-Ware™ Standard Three-Valve Delivery Cap and Parts


Parts and accessories for standard three-valve cap system

Manufacturer: kimble™  9539030004

 View more versions of this product

Catalog No. K953903-0004

  • / Each
 

Description & Specifications

Specifications

Type Valve handle
For Use With (Equipment) Ultra-ware™ standard three valve cap system
Color Yellow
No. per Case 1 Cs.