DWK Life Sciences Kimble™ Kontes™ HPLC Ultra-Ware™ Standard Three-Valve Delivery Cap and Parts
Parts and accessories for standard three-valve cap system
$33.85 - $1335.96
Specifications
For Use With (Equipment) | Ultra-ware™ standard three valve cap system |
---|---|
No. per Case | 1 Cs. |
Catalog Number | Mfr. No. | Color | Product Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Product Type | Price | Quantity | ||||
K953903-0000
|
DWK Life Sciences
9539030000 |
Blue | Three-valve Delivery Cap |
Each for $1,335.96
|
|
||||
K953903-0002
|
DWK Life Sciences
9539030002 |
Blue | Valve Handle |
Each for $33.85
|
|
||||
K953903-0005
|
DWK Life Sciences
9539030005 |
Blue | Screw Cap |
Each for $77.45
|
|
||||
K953903-0042
|
DWK Life Sciences
9539030042 |
Natural | Stem Valve |
Each for $59.79
|
|
||||
K953903-0003
|
DWK Life Sciences
9539030003 |
Red | Valve Handle |
Each for $33.85
|
|
||||
K953903-0004
|
DWK Life Sciences
9539030004 |
Yellow | Valve Handle |
Each for $33.85
|
|
||||
Description
- Three on/off CTFE valves have 1/4-28 threads top and bottom
- Valve body is PTFE
- Valve stem is CTFE
- O-ring for valve stem and lower seal is TFE/propylene
Specifications
Ultra-ware™ standard three valve cap system | |
1 Cs. |