DWK Life Sciences Kimble™ Kontes™ HPLC Ultra-Ware™ Standard Three-Valve Delivery Cap and Parts
Parts and accessories for standard three-valve cap system
Supplier: DWK Life Sciences 9539030000
Description
- Three on/off CTFE valves have 1/4-28 threads top and bottom
- Valve body is PTFE
- Valve stem is CTFE
- O-ring for valve stem and lower seal is TFE/propylene
Specifications
Three-valve Delivery Cap | |
Blue | |
CTFE | |
GL 45 |
Ultra-ware™ standard three valve cap system | |
Three on/off CTFE valve, CTFE valve stem, TFE/Propylene lower seal | |
1 Cs. |