Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Hollow White Rubber Sleeve Stoppers


Manufacturer: kimble™  7742610014

 View more versions of this product

Catalog No. K774261-0014

  • / Case of 50 Each
 

Description & Specifications

Specifications

Diameter (English) Outside 0.55 in.
Diameter (Metric) Outside 1.4cm
For Use With (Equipment) NMR tubes, pressure venting or adding reactants via syringe
Standard Taper Joint 14/20
Taper Type 14/20
Color White
Material Rubber
Product Type Plug Type Sleeve Stopper