Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Heavy Wall Recovery Flasks with Standard Bead, standard taper Outer Joint
Manufacturer: kimble™  2943000010
 View more versions of this product
Catalog No. K294300-0010
  • / Each
Description & Specifications
Specifications
For Use With (Application) Designed for easy recovery of reaction products
Capacity (Metric) 10mL
Neck Style Single
Total Count 1.0
Material Glass
Standard Taper Joint Side 14/20
Diameter (English) Outer 1.22 in.
For Use With (Equipment) Rotary evaporator
Standard Taper Joint 14/20
Diameter (Metric) Outer 31mm