Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Heavy Wall Recovery Flasks with Standard Bead, standard taper Outer Joint


Manufacturer: kimble™  6086750224

 View more versions of this product

Catalog No. K608675-0224

  • / Each
 

Description & Specifications

Specifications

Diameter (Metric) Outer 60mm
Standard Taper Joint 24/25
For Use With (Equipment) Rotary evaporator
Material Glass
Total Count 1.0
Standard Taper Joint Side 24/25
Neck Style Single
For Use With (Application) Designed for easy recovery of reaction products
Capacity (Metric) 100mL
Diameter (English) Outer 2.36 in.