Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Heavy Wall Recovery Flasks with Square Bead, standard taper Outer Joint


Ideal for use with rotary evaporators

Manufacturer: kimble™  6086756229

 View more versions of this product

Catalog No. K608675-6229

  • / Each
 

Description & Specifications

Specifications

Capacity (Metric) 100mL
Diameter (English) Outer 2.36 in.
Diameter (Metric) Outer 60mm
For Use With (Application) Square bead joints provide superior clamping to standard Joint beads
For Use With (Equipment) Rotary evaporator
Neck Style Single
Standard Taper Joint Side 29/42
Standard Taper Joint 29/42
Total Count 1.0
Material Borosilicate Glass